siRNA Details
Gene Symbol: TARDBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007375.3 | NP_031401.1 | 6 | 3756 | ||
XM_017000863.1 | XP_016856352.1 | 6 | 3744 | |||
XM_017000866.1 | XP_016856355.1 | 7 | 3010 | |||
GenBank mRNA | AF311304.1 | - | 1 | 659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
