siRNA Details
Gene Symbol: GLT8D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316967.1 | NP_001303896.1 | 6 | 1017 | ||
NM_031302.4 | NP_112592.1 | 7 | 1152 | |||
XM_011538793.2 | XP_011537095.1 | 7 | 1387 | |||
XM_017019999.1 | XP_016875488.1 | 8 | 1468 | |||
GenBank mRNA | AJ271711.1 | - | 4 | 366 | ||
AK027310.1 | - | 5 | 466 | |||
AK056056.1 | - | 6 | 1023 | |||
AY358763.1 | - | 6 | 556 | |||
BC022343.1 | - | 6 | 773 | 4248949 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
