siRNA Details
Gene Symbol: ARHGAP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031305.2 | NP_112595.2 | 1, 2 | 778 | ||
GenBank mRNA | AK308019.1 | - | 1, 2 | 123 | ||
AL136646.1 | - | 1, 2 | 780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
