siRNA Details
Gene Symbol: PGPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300927.1 | NP_001287856.1 | 4 | 6120 | ||
NM_001308366.1 | NP_001295295.1 | 5 | 6313 | |||
NM_017712.3 | NP_060182.1 | 5 | 6353 | |||
XM_006722783.3 | XP_006722846.1 | 5 | 6253 | |||
XM_011528115.1 | XP_011526417.1 | 4 | 6278 | |||
XM_017026913.1 | XP_016882402.1 | 4 | 6203 | |||
GenBank mRNA | AK025948.1 | - | 1 | 891 | ||
BC004942.1 | - | 1 | 264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
