siRNA Details
Gene Symbol: FBXW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267585.1 | NP_001254514.1 | 8 | 1774 | ||
NM_001267586.1 | NP_001254515.1 | 8 | 1774 | |||
NR_051988.1 | 8 | 2420 | ||||
XM_011523625.1 | XP_011521927.1 | 7 | 1927 | |||
XM_011523626.2 | XP_011521928.1 | 7 | 1927 | |||
XM_017024020.1 | XP_016879509.1 | 7 | 1927 | |||
XM_017024021.1 | XP_016879510.1 | 7 | 1927 | |||
GenBank mRNA | AK310383.1 | - | 8 | 2359 | ||
AL136822.1 | - | 8 | 1712 | |||
AY729024.1 | - | 8 | 1542 | |||
BC028364.2 | - | 7 | 1848 | 4827344 |
Gene Symbol: CDRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282540.1 | NP_001269469.1 | 8 | 1734 | ||
NM_006382.3 | NP_006373.2 | 8 | 1734 | |||
GenBank mRNA | AK057407.1 | - | 7 | 1562 | ||
BC143291.1 | - | 1 | 74 | |||
BC171723.1 | - | 1 | 74 | |||
U43383.1 | - | 1 | 35 | |||
U65652.1 | - | 1 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
