siRNA Details
Gene Symbol: TRPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033678.3 | NP_001028850.2 | 4, 5 | 549 | ||
NM_001160390.1 | NP_001153862.1 | 4, 5 | 537 | |||
NM_001160392.1 | NP_001153864.1 | 4, 5 | 549 | |||
NM_001160393.1 | NP_001153865.1 | 3, 4 | 344 | |||
NM_031472.3 | NP_113660.1 | 3, 4 | 465 | |||
XM_005274345.4 | XP_005274402.1 | 4, 5 | 469 | |||
XM_005274346.4 | XP_005274403.1 | 4, 5 | 428 | |||
XM_005274347.4 | XP_005274404.1 | 3, 4 | 345 | |||
XM_006718706.3 | XP_006718769.1 | 4, 5 | 431 | |||
XM_006718707.3 | XP_006718770.1 | 4, 5 | 378 | |||
XM_017018400.1 | XP_016873889.1 | 3, 4 | 345 | |||
XM_017018401.1 | XP_016873890.1 | 3, 4 | 420 | |||
GenBank mRNA | AK026364.1 | - | 3, 4 | 1058 | ||
AY211494.1 | - | 4, 5 | 505 | |||
BC005133.2 | - | 3, 4 | 361 | 3836683 | ||
BG105545.1 | - | 4, 5 | 481 | |||
BQ672032.1 | - | 3, 4 | 363 | |||
BQ674339.1 | - | 3, 4 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
