siRNA Details
Gene Symbol: ARHGAP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025616.2 | NP_001020787.2 | 5 | 871 | ||
NM_001042669.1 | NP_001036134.1 | 3 | 422 | |||
NM_001287805.1 | NP_001274734.1 | 3 | 237 | |||
NM_031305.2 | NP_112595.2 | 2 | 801 | |||
XM_011532300.2 | XP_011530602.1 | 3 | 5328 | |||
GenBank mRNA | AK098193.1 | - | 3 | 334 | ||
AK307423.1 | - | 3 | 237 | |||
AK308019.1 | - | 2 | 146 | |||
AL136646.1 | - | 2 | 803 | |||
BC047918.1 | - | 5 | 764 | |||
BC098580.1 | - | 3 | 414 | 30923119 | ||
DA546029.1 | - | 3 | 422 | |||
DA815489.1 | - | 5 | 529 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
