siRNA Details
Gene Symbol: CECR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290046.1 | NP_001276975.1 | 2 | 152 | ||
NM_001290047.1 | NP_001276976.1 | 2 | 621 | |||
XM_011546128.2 | XP_011544430.1 | 2 | 309 | |||
XM_011546129.2 | XP_011544431.1 | 2 | 309 | |||
XM_011546132.2 | XP_011544434.1 | 2 | 309 | |||
XM_017028785.1 | XP_016884274.1 | 2 | 309 | |||
GenBank mRNA | AF336133.1 | - | 2 | 621 | ||
AK308330.1 | - | 2 | 154 | |||
AL832377.1 | - | 2 | 99 | |||
BX647449.1 | - | 2 | 99 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
