siRNA Details
Gene Symbol: RASSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182663.3 | NP_872604.1 | 4, 5 | 1059 | ||
NM_182665.3 | NP_872606.1 | 3, 4 | 937 | |||
GenBank mRNA | AF445801.1 | - | 3, 4 | 937 | ||
AK074115.1 | - | 2, 3 | 404 | |||
AK289861.1 | - | 4, 5 | 1059 | |||
AK291268.1 | - | 4, 5 | 1057 | |||
AK292039.1 | - | 4, 5 | 795 | |||
AL832784.1 | - | 3, 4 | 837 | |||
AY062002.1 | - | 4, 5 | 1043 | |||
AY216268.1 | - | 3, 4 | 930 | |||
AY261332.1 | - | 3, 4 | 837 | |||
BC004270.1 | - | 4, 5 | 1043 | 3613993 | ||
BC007203.1 | - | 4, 5 | 1043 | 2967057 | ||
BC042651.1 | - | 3, 4 | 917 | 4342962 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
