siRNA Details
Gene Symbol: FBXW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267585.1 | NP_001254514.1 | 8, 9 | 1788 | ||
NM_001267586.1 | NP_001254515.1 | 8, 9 | 1788 | |||
NR_051988.1 | 8, 9 | 2434 | ||||
XM_011523625.1 | XP_011521927.1 | 7, 8 | 1941 | |||
XM_011523626.2 | XP_011521928.1 | 7, 8 | 1941 | |||
XM_017024020.1 | XP_016879509.1 | 7, 8 | 1941 | |||
XM_017024021.1 | XP_016879510.1 | 7, 8 | 1941 | |||
GenBank mRNA | AK310383.1 | - | 8, 9 | 2373 | ||
AL136822.1 | - | 8, 9 | 1726 | |||
AY729024.1 | - | 8, 9 | 1556 | |||
BC028364.2 | - | 7, 8 | 1862 | 4827344 |
Gene Symbol: CDRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282540.1 | NP_001269469.1 | 8, 9 | 1748 | ||
NM_006382.3 | NP_006373.2 | 8, 9 | 1748 | |||
GenBank mRNA | AK057407.1 | - | 7, 8 | 1576 | ||
BC143291.1 | - | 1, 2 | 88 | |||
BC171723.1 | - | 1, 2 | 88 | |||
U43383.1 | - | 1, 2 | 49 | |||
U65652.1 | - | 1, 2 | 277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
