siRNA Details
Gene Symbol: CD99L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184808.1 | NP_001171737.1 | 3, 4 | 438 | ||
NM_031462.3 | NP_113650.2 | 3, 4 | 438 | |||
XM_017029890.1 | XP_016885379.1 | 3, 4 | 302 | |||
GenBank mRNA | AK023776.1 | - | 2, 3 | 124 | ||
AK290200.1 | - | 3, 4 | 214 | |||
AK293244.1 | - | 3, 4 | 244 | |||
AK295217.1 | - | 3, 4 | 256 | |||
AL136580.1 | - | 3, 4 | 328 | |||
AY078165.1 | - | 3, 4 | 246 | |||
AY358837.1 | - | 3, 4 | 311 | |||
BC007569.1 | - | 2, 3 | 98 | |||
BC013029.2 | - | 2, 3 | 98 | |||
BC025729.1 | - | 4, 5 | 1551 | 5203087 | ||
BC030536.1 | - | 3, 4 | 298 | 5242536 | ||
DC323066.1 | - | 3, 4 | 438 | |||
KU178797.1 | - | 3, 4 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
