siRNA Details
Gene Symbol: TRAPPC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160372.3 | NP_001153844.1 | 23 | 4066 | ||
NM_001321646.1 | NP_001308575.1 | 22 | 4039 | |||
NM_031466.7 | NP_113654.4 | 23 | 4241 | |||
XM_011517326.2 | XP_011515628.1 | 22 | 4214 | |||
XM_011517330.2 | XP_011515632.1 | 15 | 2606 | |||
XM_017013894.1 | XP_016869383.1 | 16 | 2942 | |||
GenBank mRNA | AB067469.1 | - | 23 | 4016 | ||
AK026394.1 | - | 23 | 4058 | |||
AK027689.1 | - | 12 | 2166 | |||
AK128755.1 | - | 23 | 4242 | |||
AL833973.1 | - | 21 | 3341 | |||
AY630619.1 | - | 4 | 1121 | |||
BC006206.2 | - | 21 | 3462 | 3538806 | ||
BC065288.1 | - | 6 | 1326 | |||
BP394939.1 | - | 1 | 87 |
