siRNA Details
Gene Symbol: SH3KBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031892.2 | NP_114098.1 | 2 | 460 | ||
XM_005274494.2 | XP_005274551.1 | 2 | 326 | |||
XM_011545498.2 | XP_011543800.1 | 2 | 326 | |||
XM_011545500.2 | XP_011543802.1 | 2 | 326 | |||
XM_011545503.2 | XP_011543805.1 | 2 | 326 | |||
XM_017029462.1 | XP_016884951.1 | 2 | 326 | |||
XM_017029466.1 | XP_016884955.1 | 2 | 326 | |||
GenBank mRNA | AF230904.1 | - | 2 | 435 | ||
AK313308.1 | - | 2 | 388 | |||
AY423734.1 | - | 3 | 358 | |||
AY542305.1 | - | 2 | 195 | |||
BC015806.1 | - | 2 | 362 | 3906722 | ||
BC050663.2 | - | 2 | 370 | |||
DR000949.1 | - | 2 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
