siRNA Details
Gene Symbol: PRPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002764.3 | NP_002755.1 | 1, 2 | 327 | ||
GenBank mRNA | AK223169.1 | - | 1, 2 | 144 | ||
AK297968.1 | - | 2, 3 | 165 | |||
AK307748.1 | - | 1, 2 | 241 | |||
AK312706.1 | - | 1, 2 | 241 | |||
BC001605.1 | - | 1, 2 | 222 | 3542584 | ||
D00860.1 | - | 1, 2 | 244 | |||
DC411129.1 | - | 1, 2 | 323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
