siRNA Details
Gene Symbol: PRPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002764.3 | NP_002755.1 | 2 | 366 | ||
GenBank mRNA | AK223169.1 | - | 2 | 183 | ||
AK297968.1 | - | 3 | 204 | |||
AK307748.1 | - | 2 | 280 | |||
AK312706.1 | - | 2 | 280 | |||
BC001605.1 | - | 2 | 261 | 3542584 | ||
D00860.1 | - | 2 | 283 | |||
DC411129.1 | - | 2 | 362 | |||
X15331.1 | - | 2 | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
