siRNA Details
Gene Symbol: CCDC93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019044.4 | NP_061917.3 | 24 | 6032 | ||
XM_011511359.1 | XP_011509661.1 | 24 | 6029 | |||
XM_011511361.1 | XP_011509663.1 | 23 | 5786 | |||
XM_011511362.1 | XP_011509664.1 | 23 | 5900 | |||
GenBank mRNA | AB209993.1 | - | 24 | 5950 | ||
AF070582.1 | - | 1 | 943 | |||
AK124683.1 | - | 1 | 2076 | |||
AL050001.1 | - | 1 | 332 | |||
AY726577.1 | - | 1 | 1996 | |||
BC005078.1 | - | 1 | 520 | |||
BC080559.1 | - | 1 | 502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
