siRNA Details
Gene Symbol: CDRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282540.1 | NP_001269469.1 | 9 | 1822 | ||
NM_006382.3 | NP_006373.2 | 9 | 1822 | |||
GenBank mRNA | AK057407.1 | - | 8 | 1650 | ||
AW891009.1 | - | 1 | 56 | |||
BC143291.1 | - | 2 | 162 | |||
BC171723.1 | - | 2 | 162 | |||
U43383.1 | - | 2 | 123 | |||
U65652.1 | - | 2 | 351 |
Gene Symbol: FBXW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267585.1 | NP_001254514.1 | 9 | 1862 | ||
NM_001267586.1 | NP_001254515.1 | 9 | 1862 | |||
NR_051988.1 | 9 | 2508 | ||||
XM_011523625.1 | XP_011521927.1 | 8 | 2015 | |||
XM_011523626.2 | XP_011521928.1 | 8 | 2015 | |||
XM_017024020.1 | XP_016879509.1 | 8 | 2015 | |||
XM_017024021.1 | XP_016879510.1 | 8 | 2015 | |||
GenBank mRNA | AK310383.1 | - | 9 | 2447 | ||
AL136822.1 | - | 9 | 1800 | |||
AY729024.1 | - | 9 | 1630 | |||
BC028364.2 | - | 8 | 1936 | 4827344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
