siRNA Details
Gene Symbol: SCML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006089.2 | NP_006080.1 | 15 | 2975 | ||
XM_006724458.1 | XP_006724521.1 | 14 | 2787 | |||
XM_017029221.1 | XP_016884710.1 | 14 | 2857 | |||
GenBank mRNA | AK022824.1 | - | 3 | 1223 | ||
BC064617.1 | - | 15 | 2932 | 5575894 | ||
Y18004.1 | - | 15 | 2908 |
