siRNA Details
Gene Symbol: NDRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032013.3 | NP_114402.1 | 3, 4 | 228 | ||
XM_006723837.3 | XP_006723900.1 | 3, 4 | 171 | |||
XM_017027978.1 | XP_016883467.1 | 3, 4 | 468 | |||
XM_017027979.1 | XP_016883468.1 | 3, 4 | 468 | |||
GenBank mRNA | AB044943.1 | - | 3, 4 | 168 | ||
AK023618.1 | - | 3, 4 | 168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
