siRNA Details
Gene Symbol: RACGAP1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026583.1 | 1 | 137 | |||
GenBank mRNA | AF334184.1 | - | 1 | 137 | ||
BC125190.1 | - | 1 | 128 | |||
BC125191.1 | - | 1 | 128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
