siRNA Details
Gene Symbol: LTBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000428.2 | NP_000419.1 | 36 | 8406 | ||
XM_011536765.1 | XP_011535067.1 | 34 | 7933 | |||
GenBank mRNA | AB209865.1 | - | 36 | 7656 | ||
AF113211.1 | - | 1 | 1120 | |||
BC062686.1 | - | 1 | 913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
