siRNA Details
Gene Symbol: B3GNT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032047.4 | NP_114436.1 | 2 | 778 | ||
XM_005247823.4 | XP_005247880.1 | 2 | 983 | |||
XM_005247824.3 | XP_005247881.1 | 3 | 1037 | |||
XM_005247825.3 | XP_005247882.1 | 3 | 921 | |||
XM_011513225.2 | XP_011511527.1 | 2 | 1417 | |||
XM_011513226.2 | XP_011511528.1 | 3 | 894 | |||
XM_011513227.2 | XP_011511529.1 | 3 | 928 | |||
XM_011513228.2 | XP_011511530.1 | 2 | 829 | |||
XM_011513229.2 | XP_011511531.1 | 2 | 683 | |||
XM_017007309.1 | XP_016862798.1 | 3 | 2051 | |||
GenBank mRNA | AB045278.1 | - | 1 | 377 | ||
AB209517.1 | - | 2 | 596 | |||
AF368169.1 | - | 1 | 708 | |||
AJ304505.1 | - | 1 | 291 | |||
AK074424.1 | - | 2 | 765 | |||
AK091474.1 | - | 2 | 965 | |||
BC028058.1 | - | 2 | 771 | 5175670 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
