siRNA Details
Gene Symbol: NACAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002182.1 | 1 | 544 | |||
GenBank mRNA | AF315951.1 | - | 2 | 606 | ||
AK291701.1 | - | 3 | 1076 | |||
BC069062.1 | - | 1 | 544 | |||
BC069411.1 | - | 1 | 637 | |||
BC069542.1 | - | 1 | 632 | |||
BC074958.2 | - | 1 | 632 | |||
BC074959.2 | - | 1 | 632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
