siRNA Details
Gene Symbol: C10orf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305581.1 | NP_001292510.1 | 4 | 459 | ||
NM_032024.4 | NP_114413.1 | 3 | 511 | |||
NR_131178.1 | 5 | 734 | ||||
XM_011540256.2 | XP_011538558.1 | 4 | 453 | |||
XM_011540257.2 | XP_011538559.1 | 4 | 453 | |||
XM_017016759.1 | XP_016872248.1 | 4 | 453 | |||
XM_017016760.1 | XP_016872249.1 | 4 | 453 | |||
XM_017016761.1 | XP_016872250.1 | 4 | 453 | |||
GenBank mRNA | AF267860.1 | - | 3 | 512 | ||
BM720401.1 | - | 3 | 459 | |||
BX105111.1 | - | 3 | 352 | |||
CR933639.1 | - | 5 | 734 | |||
HY006640.1 | - | 4 | 450 |
