siRNA Details
Gene Symbol: KIRREL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032123.6 | NP_115499.5 | 16 | 2053 | ||
NM_199179.3 | NP_954648.3 | 15 | 1903 | |||
NM_199180.3 | NP_954649.3 | 15 | 2464 | |||
XM_011527362.1 | XP_011525664.1 | 16 | 2524 | |||
XM_011527363.1 | XP_011525665.1 | 16 | 2505 | |||
XM_011527365.1 | XP_011525667.1 | 15 | 2374 | |||
XM_011527366.1 | XP_011525668.1 | 14 | 2635 | |||
XM_017027348.1 | XP_016882837.1 | 14 | 2926 | |||
GenBank mRNA | AB593116.1 | - | 15 | 2428 | ||
AL136654.1 | - | 15 | 2448 | |||
AY358742.1 | - | 15 | 2450 | |||
BC007312.1 | - | 6 | 997 | |||
BC064925.1 | - | 18 | 2387 | 6141879 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
