siRNA Details
Gene Symbol: SLC10A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317817.1 | NP_001304746.1 | 4 | 936 | ||
NM_032128.4 | NP_115504.1 | 5 | 1073 | |||
GenBank mRNA | AL136728.1 | - | 5 | 1001 | ||
BC063471.1 | - | 5 | 986 | |||
BC120970.1 | - | 5 | 932 | |||
BC120971.1 | - | 4 | 795 | |||
BC127626.1 | - | 5 | 932 | |||
BC127627.1 | - | 5 | 932 | |||
BC148252.1 | - | 5 | 932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
