siRNA Details
Gene Symbol: ARMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290020.1 | NP_001276949.1 | 14 | 2168 | ||
NM_001290021.1 | NP_001276950.1 | 7 | 994 | |||
NM_001312689.1 | NP_001299618.1 | 9 | 1385 | |||
NM_018076.4 | NP_060546.2 | 14 | 2121 | |||
XM_011519527.1 | XP_011517829.1 | 14 | 2168 | |||
XM_011519528.1 | XP_011517830.1 | 14 | 2818 | |||
XM_011519533.2 | XP_011517835.1 | 9 | 1342 | |||
XM_011519534.2 | XP_011517836.1 | 9 | 1475 | |||
XM_011519535.2 | XP_011517837.1 | 8 | 1227 | |||
XM_011519537.2 | XP_011517839.1 | 5 | 902 | |||
XM_017016368.1 | XP_016871857.1 | 14 | 2168 | |||
XM_017016369.1 | XP_016871858.1 | 14 | 2168 | |||
XM_017016370.1 | XP_016871859.1 | 13 | 1958 | |||
XM_017016371.1 | XP_016871860.1 | 14 | 2168 | |||
GenBank mRNA | AK001679.1 | - | 14 | 2121 | ||
AK292521.1 | - | 14 | 2124 | |||
AK302048.1 | - | 7 | 948 | |||
AK302419.1 | - | 9 | 1717 | |||
AK302627.1 | - | 9 | 1385 | |||
AL136859.1 | - | 3 | 507 | |||
BC140846.1 | - | 13 | 2049 | 9021714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
