siRNA Details
Gene Symbol: PRAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032152.4 | NP_115528.4 | 3, 4 | 1571 | ||
XM_005272502.2 | XP_005272559.1 | 3, 4 | 1557 | |||
XM_011528352.2 | XP_011526654.1 | 3, 4 | 1577 | |||
XM_011528353.2 | XP_011526655.1 | 3, 4 | 1575 | |||
GenBank mRNA | AJ272324.1 | - | 2, 3 | 1715 | ||
AK308227.1 | - | 3, 4 | 1554 | |||
BC028012.1 | - | 4, 5 | 1528 | 5215850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
