siRNA Details
Gene Symbol: MUM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032853.3 | NP_116242.2 | 14 | 3543 | ||
NR_024247.1 | 14 | 3612 | ||||
XM_011528380.2 | XP_011526682.1 | 15 | 3635 | |||
GenBank mRNA | AK025936.1 | - | 1 | 1018 | ||
AK075241.1 | - | 8 | 2173 | |||
AL360266.1 | - | 2 | 1423 | |||
BC019585.1 | - | 1 | 527 | |||
BC021016.1 | - | 1 | 700 | |||
BC082987.1 | - | 5 | 1994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
