siRNA Details
Gene Symbol: ELL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006532.3 | NP_006523.1 | 12 | 3270 | ||
XM_017027335.1 | XP_016882824.1 | 13 | 3254 | |||
XM_017027336.1 | XP_016882825.1 | 8 | 3006 | |||
XM_017027337.1 | XP_016882826.1 | 7 | 2446 | |||
GenBank mRNA | AB208794.1 | - | 13 | 3251 | ||
AF157562.1 | - | 1 | 207 | |||
AL136771.1 | - | 1 | 240 | |||
BC064558.1 | - | 1 | 328 | |||
BC108256.1 | - | 1 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
