siRNA Details
Gene Symbol: CYP4F30P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023391.1 | 1 | 621 | |||
GenBank mRNA | AL136789.1 | - | 1 | 621 |
Gene Symbol: CYP4F62P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103761.1 | 1 | 827 |
Gene Symbol: FAR2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026758.2 | 1 | 1136 | |||
GenBank mRNA | AK125921.1 | - | 1 | 1138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
