siRNA Details
Gene Symbol: PPIL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111298.2 | NP_001104768.2 | 9 | 2012 | ||
NM_001286360.1 | NP_001273289.1 | 7 | 1838 | |||
NM_001286361.1 | NP_001273290.1 | 6 | 1702 | |||
NM_173672.4 | NP_775943.1 | 8 | 1934 | |||
NR_104429.1 | 9 | 2054 |
Gene Symbol: MRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031727.2 | NP_001026897.1 | 6 | 1500 | ||
NM_032285.2 | NP_115661.1 | 6 | 1359 | |||
XM_011528355.2 | XP_011526657.1 | 6 | 1435 | |||
GenBank mRNA | AL834276.1 | - | 6 | 1350 | ||
BC001703.1 | - | 6 | 1474 | 3501899 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
