siRNA Details
Gene Symbol: GTF2IRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095257.1 | - | 13 | 1264 | ||
AK225845.1 | - | 16 | 1661 | |||
AL834153.1 | - | 3 | 370 | |||
AY260739.1 | - | 16 | 1657 | |||
AY312853.1 | - | 16 | 1657 | |||
AY312854.1 | - | 15 | 1497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
