siRNA Details
Gene Symbol: PHF20L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277196.1 | NP_001264125.1 | 19 | 3137 | ||
NM_016018.4 | NP_057102.4 | 20 | 3215 | |||
XM_005250931.1 | XP_005250988.1 | 20 | 3200 | |||
XM_011517075.1 | XP_011515377.1 | 20 | 3203 | |||
XM_011517076.1 | XP_011515378.1 | 20 | 3199 | |||
XM_011517077.1 | XP_011515379.1 | 20 | 3200 | |||
XM_011517078.1 | XP_011515380.1 | 20 | 3191 | |||
XM_011517079.1 | XP_011515381.1 | 20 | 3188 | |||
XM_011517080.1 | XP_011515382.1 | 19 | 3113 | |||
XM_011517081.1 | XP_011515383.1 | 19 | 3110 | |||
XM_017013509.1 | XP_016868998.1 | 20 | 3184 | |||
XM_017013510.1 | XP_016868999.1 | 19 | 3109 | |||
XM_017013511.1 | XP_016869000.1 | 19 | 3106 | |||
XM_017013512.1 | XP_016869001.1 | 19 | 3103 | |||
XM_017013513.1 | XP_016869002.1 | 20 | 6026 | |||
XM_017013514.1 | XP_016869003.1 | 20 | 6022 | |||
XM_017013515.1 | XP_016869004.1 | 13 | 2657 | |||
GenBank mRNA | AK025268.1 | - | 3 | 630 | ||
AK057961.1 | - | 7 | 1229 | |||
AK126085.1 | - | 2 | 1465 | |||
AY279109.1 | - | 7 | 1229 | |||
BC050655.1 | - | 6 | 1054 | |||
BC056413.1 | - | 3 | 682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
