siRNA Details
Gene Symbol: TET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540204.2 | XP_011538506.1 | 13 | 14284 | ||
XM_011540205.2 | XP_011538507.1 | 12 | 11927 | |||
XM_011540207.2 | XP_011538509.1 | 12 | 12164 | |||
XM_017016686.1 | XP_016872175.1 | 11 | 11968 | |||
XM_017016687.1 | XP_016872176.1 | 13 | 12218 | |||
XM_017016688.1 | XP_016872177.1 | 13 | 12098 | |||
XM_017016689.1 | XP_016872178.1 | 12 | 12048 |
Gene Symbol: ANKRD26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015929.1 | XP_016871418.1 | 41 | 13579 |
Gene Symbol: ANXA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209868.1 | - | 1 | 289 |
Gene Symbol: ATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011533018.2 | XP_011531320.1 | 1 | 234 |
Gene Symbol: DTWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308081.1 | NP_001295010.1 | 6 | 5342 | ||
NM_173666.3 | NP_775937.1 | 6 | 5563 | |||
XM_011543338.2 | XP_011541640.2 | 7 | 6956 | |||
XM_011543340.2 | XP_011541642.1 | 7 | 5455 |
Gene Symbol: FBXL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184906.1 | NP_001171835.1 | 14 | 5839 | ||
NM_032875.2 | NP_116264.2 | 15 | 5935 | |||
XM_005257746.3 | XP_005257803.2 | 15 | 6126 | |||
XM_005257747.3 | XP_005257804.2 | 14 | 5907 | |||
GenBank mRNA | AK024690.1 | - | 1 | 1575 |
Gene Symbol: FLJ13773 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023835.1 | - | 1 | 3025 |
Gene Symbol: GOLGA2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124885.1 | - | 1 | 3355 |
Gene Symbol: GRAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011523857.2 | XP_011522159.1 | 5 | 5665 |
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000202.7 | NP_000193.1 | 9 | 5492 | ||
NM_001166550.3 | NP_001160022.1 | 9 | 5448 | |||
GenBank mRNA | AK055600.1 | - | 1 | 2883 | ||
AK057191.1 | - | 1 | 2054 | |||
AL117536.1 | - | 1 | 1392 | |||
BX647357.1 | - | 9 | 5345 |
Gene Symbol: IVD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159508.1 | NP_001152980.1 | 11 | 2983 | ||
NM_002225.3 | NP_002216.2 | 12 | 3073 | |||
XM_017022152.1 | XP_016877641.1 | 12 | 2854 | |||
XM_017022154.1 | XP_016877643.1 | 12 | 2995 | |||
XM_017022155.1 | XP_016877644.1 | 12 | 1805 | |||
XM_017022156.1 | XP_016877645.1 | 12 | 3140 | |||
GenBank mRNA | AK092168.1 | - | 2 | 384 | ||
AK122922.1 | - | 11 | 2673 | |||
AY826821.1 | - | 1 | 767 | |||
AY826822.1 | - | 1 | 767 | |||
AY826823.1 | - | 1 | 767 |
Gene Symbol: KCMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020122.4 | NP_064507.3 | 7 | 6375 |
Gene Symbol: LINC00664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040866.1 | - | 2 | 1522 |
Gene Symbol: MAP3K14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123696.1 | - | 2 | 1159 |
Gene Symbol: PCNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB023212.1 | - | 10 | 4336 |
Gene Symbol: RPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712677.3 | XP_006712740.1 | 7 | 6407 |
Gene Symbol: ZNF782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC020163.1 | - | 1 | 3419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
