siRNA Details
Gene Symbol: TBC1D3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291464.1 | NP_001278393.1 | 7 | 504 | ||
XM_006722235.2 | XP_006722298.1 | 6 | 2782 | |||
XM_006722236.1 | XP_006722299.1 | 7 | 1354 | |||
XM_006722237.2 | XP_006722300.1 | 6 | 2599 | |||
XM_006722239.2 | XP_006722302.1 | 6 | 2599 | |||
XM_011524172.1 | XP_011522474.1 | 6 | 2782 | |||
XM_011524173.1 | XP_011522475.1 | 6 | 2782 | |||
XM_011524174.1 | XP_011522476.1 | 6 | 2782 |
Gene Symbol: TBC1D3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291466.1 | NP_001278395.1 | 7 | 502 | ||
XM_006722254.1 | XP_006722317.1 | 10 | 851 | |||
GenBank mRNA | AL136860.1 | - | 7 | 502 |
Gene Symbol: TBC1D3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DC398423.1 | - | 7 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
