siRNA Details
Gene Symbol: MRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031727.2 | NP_001026897.1 | 6 | 1841 | ||
NM_032285.2 | NP_115661.1 | 6 | 1700 | |||
XM_011528355.2 | XP_011526657.1 | 6 | 1776 | |||
GenBank mRNA | AL834276.1 | - | 6 | 1691 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
