siRNA Details
Gene Symbol: ARMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290020.1 | NP_001276949.1 | 20 | 3270 | ||
NM_001290021.1 | NP_001276950.1 | 14 | 2218 | |||
NM_001312689.1 | NP_001299618.1 | 15 | 2487 | |||
NM_018076.4 | NP_060546.2 | 20 | 3223 | |||
XM_011519527.1 | XP_011517829.1 | 21 | 3330 | |||
XM_011519528.1 | XP_011517830.1 | 21 | 3980 | |||
XM_011519533.2 | XP_011517835.1 | 16 | 2504 | |||
XM_011519534.2 | XP_011517836.1 | 16 | 2637 | |||
XM_011519535.2 | XP_011517837.1 | 15 | 2389 | |||
XM_011519537.2 | XP_011517839.1 | 12 | 2064 | |||
XM_017016370.1 | XP_016871859.1 | 20 | 3120 | |||
GenBank mRNA | AK001679.1 | - | 20 | 3223 | ||
AK292521.1 | - | 20 | 3226 | |||
AK302048.1 | - | 14 | 2172 | |||
AK302627.1 | - | 15 | 2487 | |||
AL136859.1 | - | 9 | 1609 | |||
BC140846.1 | - | 19 | 3151 | 9021714 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
