siRNA Details
Gene Symbol: PCGF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011663.1 | NP_001011663.1 | 4, 5 | 685 | ||
GenBank mRNA | AB047006.1 | - | 4, 5 | 639 | ||
AK290679.1 | - | 4, 5 | 639 | |||
AK309105.1 | - | 4, 5 | 707 | |||
BC010235.1 | - | 4, 5 | 691 | 3461513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
