siRNA Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201269.2 | NP_958357.1 | 4 | 3727 | ||
XM_005271257.4 | XP_005271314.1 | 5 | 4063 | |||
XM_005271260.4 | XP_005271317.1 | 4 | 4040 | |||
XM_011542258.2 | XP_011540560.1 | 4 | 3741 | |||
XM_011542259.2 | XP_011540561.1 | 6 | 4417 | |||
XM_011542260.2 | XP_011540562.1 | 6 | 4321 | |||
XM_011542261.2 | XP_011540563.1 | 4 | 3662 | |||
XM_017002487.1 | XP_016857976.1 | 5 | 3852 | |||
XM_017002488.1 | XP_016857977.1 | 5 | 4063 | |||
XM_017002489.1 | XP_016857978.1 | 4 | 4038 | |||
XM_017002490.1 | XP_016857979.1 | 5 | 3850 | |||
XM_017002491.1 | XP_016857980.1 | 4 | 3662 | |||
GenBank mRNA | AB019255.1 | - | 3466 | |||
AB033047.1 | - | 2 | 3310 | |||
AK001587.1 | - | 2 | 1492 | |||
AK023596.1 | - | 2 | 666 | |||
BC031003.1 | - | 1 | 114 | |||
BC063683.1 | - | 3 | 3421 | |||
BC132776.1 | - | 4 | 3542 | 40146798 | ||
BC132778.1 | - | 4 | 3542 | 40146800 | ||
BC150177.1 | - | 4 | 3682 | 40148078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
