siRNA Details
Gene Symbol: TCEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006756.3 | NP_006747.1 | 2 | 426 | ||
NR_109901.1 | 2 | 426 | ||||
NR_109902.1 | 2 | 426 | ||||
XM_006716467.1 | XP_006716530.1 | 2 | 280 | |||
XM_011517578.1 | XP_011515880.1 | 2 | 357 | |||
GenBank mRNA | AK026617.1 | - | 2 | 229 | ||
AK290454.1 | - | 2 | 113 | |||
AK297729.1 | - | 2 | 239 | |||
AU280634.1 | - | 2 | 421 | |||
BC072460.1 | - | 2 | 221 | 6470030 | ||
BM477937.1 | - | 382 | ||||
BT019995.1 | - | 1 | 95 | |||
BX538034.1 | - | 2 | 237 | |||
CR542221.1 | - | 2 | 95 | |||
DB091231.1 | - | 2 | 429 | |||
M81601.1 | - | 2 | 268 | |||
X62585.1 | - | 2 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
