siRNA Details
Gene Symbol: SETD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032233.2 | NP_115609.2 | 13 | 2505 | ||
XM_005268127.3 | XP_005268184.1 | 12 | 2358 | |||
XM_011537231.2 | XP_011535533.2 | 13 | 2457 | |||
XM_011537232.2 | XP_011535534.1 | 13 | 3635 | |||
XM_011537233.2 | XP_011535535.1 | 11 | 2283 | |||
XM_011537234.1 | XP_011535536.1 | 12 | 2225 | |||
XM_017021699.1 | XP_016877188.1 | 12 | 2353 | |||
XM_017021700.1 | XP_016877189.1 | 13 | 2541 | |||
XM_017021701.1 | XP_016877190.1 | 11 | 2333 | |||
XM_017021702.1 | XP_016877191.1 | 10 | 2152 | |||
GenBank mRNA | AK026680.1 | - | 12 | 2259 | ||
AL359581.2 | - | 8 | 1695 | |||
BC142995.1 | - | 13 | 2518 | 8860450 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
