siRNA Details
Gene Symbol: ANKRD20A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012421.1 | NP_001012421.1 | 12, 13 | 1272 | ||
XM_011517877.2 | XP_011516179.1 | 12, 13 | 1272 | |||
XM_011517878.2 | XP_011516180.1 | 12, 13 | 1272 | |||
XM_011517879.2 | XP_011516181.1 | 12, 13 | 1222 |
Gene Symbol: ANKRD20A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032250.3 | NP_115626.2 | 12, 13 | 1272 | ||
XM_005272532.4 | XP_005272589.1 | 12, 13 | 1206 | |||
XM_011545682.2 | XP_011543984.1 | 12, 13 | 1209 | |||
GenBank mRNA | AL136793.1 | - | 12, 13 | 1273 |
Gene Symbol: ANKRD20A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012419.2 | NP_001012419.1 | 12, 13 | 1238 | ||
XM_005272525.3 | XP_005272582.1 | 12, 13 | 1491 | |||
XM_011543860.2 | XP_011542162.1 | 12, 13 | 1495 | |||
XM_011543861.2 | XP_011542163.1 | 12, 13 | 1499 | |||
GenBank mRNA | BC140809.1 | - | 12, 13 | 1235 | 9021677 | |
BC171793.1 | - | 12, 13 | 1235 | 9054447 |
Gene Symbol: ANKRD20A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098805.1 | NP_001092275.1 | 12, 13 | 1272 |
Gene Symbol: ANKRD20A9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BJ991373.1 | - | 5, 6 | 467 | ||
DW423875.1 | - | 3, 4 | 210 |
Gene Symbol: LOC102723382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546266.2 | XP_011544568.1 | 4, 5 | 353 |
Gene Symbol: LOC105379417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030104.1 | XP_016885593.1 | 7, 8 | 1789 |
Gene Symbol: LOC105379426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC016022.1 | - | 9, 10 | 852 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
