siRNA Details
Gene Symbol: TBC1D3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291466.1 | NP_001278395.1 | 7 | 567 | ||
XM_006722254.1 | XP_006722317.1 | 10 | 916 | |||
GenBank mRNA | AL136860.1 | - | 7 | 567 | ||
BC047739.1 | - | 8 | 780 |
Gene Symbol: LOC101060389 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291465.1 | NP_001278394.1 | 7 | 569 | ||
XM_006722248.2 | XP_006722311.1 | 6 | 2075 | |||
XM_006722249.2 | XP_006722312.1 | 6 | 2075 | |||
XM_006722250.2 | XP_006722313.1 | 6 | 2075 | |||
XM_006722252.3 | XP_006722315.1 | 6 | 1892 | |||
XM_006722253.2 | XP_006722316.1 | 6 | 2075 | |||
XM_011524176.2 | XP_011522478.1 | 7 | 682 | |||
XM_017024010.1 | XP_016879499.1 | 6 | 2075 | |||
GenBank mRNA | BI088323.1 | - | 5 | 332 |
Gene Symbol: TBC1D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123391.3 | NP_001116863.3 | 7 | 581 | ||
GenBank mRNA | AB449884.1 | - | 6 | 469 | ||
AK302064.1 | - | 8 | 807 | |||
AK303259.1 | - | 6 | 601 | |||
AX775941.1 | - | 9 | 830 | |||
AX775943.1 | - | 9 | 830 |
Gene Symbol: TBC1D3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF540953.1 | - | 6 | 469 |
Gene Symbol: TBC1D3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001418.6 | NP_001001418.5 | 7 | 609 | ||
XM_006721904.2 | XP_006721967.1 | 6 | 2075 | |||
XM_006721905.2 | XP_006721968.1 | 6 | 2075 | |||
XM_006721907.3 | XP_006721970.1 | 6 | 1892 | |||
XM_006721908.2 | XP_006721971.1 | 6 | 2075 | |||
XM_011524814.2 | XP_011523116.1 | 7 | 682 | |||
XM_017024646.1 | XP_016880135.1 | 6 | 2075 | |||
GenBank mRNA | AK313329.1 | - | 7 | 609 |
Gene Symbol: TBC1D3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032258.4 | NP_115634.3 | 7 | 583 |
Gene Symbol: TBC1D3I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291463.1 | NP_001278392.1 | 7 | 569 | ||
XM_006722223.3 | XP_006722286.1 | 7 | 840 | |||
XM_006722224.3 | XP_006722287.1 | 6 | 2085 | |||
XM_006722226.3 | XP_006722289.1 | 6 | 2085 | |||
XM_011524183.2 | XP_011522485.1 | 6 | 2268 | |||
XM_011524184.2 | XP_011522486.1 | 6 | 2268 | |||
XM_011524185.2 | XP_011522487.1 | 6 | 2268 | |||
GenBank mRNA | BC078140.1 | - | 7 | 570 | 5742850 |
Gene Symbol: TBC1D3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025494.1 | XP_016880983.1 | 5 | 385 |
Gene Symbol: TBC1D3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291464.1 | NP_001278393.1 | 7 | 569 | ||
XM_006722235.2 | XP_006722298.1 | 6 | 2847 | |||
XM_006722236.1 | XP_006722299.1 | 7 | 1419 | |||
XM_006722237.2 | XP_006722300.1 | 6 | 2664 | |||
XM_006722239.2 | XP_006722302.1 | 6 | 2664 | |||
XM_011524172.1 | XP_011522474.1 | 6 | 2847 | |||
XM_011524173.1 | XP_011522475.1 | 6 | 2847 | |||
XM_011524174.1 | XP_011522476.1 | 6 | 2847 |
Gene Symbol: TBC1D3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291459.1 | NP_001278388.1 | 7 | 583 | ||
XM_006722242.3 | XP_006722305.1 | 6 | 3823 | |||
XM_006722243.2 | XP_006722306.1 | 6 | 2075 | |||
XM_006722244.2 | XP_006722307.1 | 6 | 2075 | |||
XM_006722246.3 | XP_006722309.1 | 6 | 1223 | |||
XM_006722247.2 | XP_006722310.1 | 6 | 2075 | |||
XM_011524175.2 | XP_011522477.1 | 7 | 682 | |||
GenBank mRNA | AK095385.1 | - | 6 | 1223 | ||
AK122833.1 | - | 3 | 1894 | |||
AX775949.1 | - | 6 | 1223 | |||
AX775951.1 | - | 6 | 1223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
