siRNA Details
Gene Symbol: ZFHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006885.3 | NP_008816.3 | 2 | 1320 | ||
XM_005255957.3 | XP_005256014.1 | 2 | 9798 | |||
GenBank mRNA | L32832.1 | - | 2 | 1320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
