siRNA Details
Gene Symbol: HSDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195822.1 | NP_001182751.1 | 9 | 1221 | ||
NM_032303.4 | NP_115679.2 | 11 | 1440 | |||
NR_036651.1 | 10 | 1276 | ||||
XM_017015203.1 | XP_016870692.1 | 8 | 1674 | |||
XM_017015204.1 | XP_016870693.1 | 10 | 1953 | |||
XM_017015205.1 | XP_016870694.1 | 9 | 1789 | |||
GenBank mRNA | AK090940.1 | - | 3 | 236 | ||
AK098721.1 | - | 11 | 1411 | |||
AK225946.1 | - | 8 | 973 | |||
AK289929.1 | - | 11 | 1268 | |||
AK292486.1 | - | 9 | 1028 | |||
AK301469.1 | - | 9 | 1034 | |||
AK303645.1 | - | 10 | 1082 | |||
AK313605.1 | - | 9 | 1028 | |||
AL833735.1 | - | 9 | 954 | |||
AY093428.1 | - | 11 | 1279 | |||
BC004331.1 | - | 9 | 1050 | 3630835 | ||
BC095451.1 | - | 11 | 1217 | 30405073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
