siRNA Details
Gene Symbol: ACBD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032360.3 | NP_115736.1 | 1, 2 | 841 | ||
GenBank mRNA | AK314112.1 | - | 1, 2 | 816 | ||
AL537685.3 | - | 1, 2 | 839 | |||
AL561590.3 | - | 1, 2 | 836 | |||
AY606133.1 | - | 1, 2 | 254 | |||
BC006505.2 | - | 1, 2 | 298 | 2821736 | ||
BU170874.1 | - | 1, 2 | 769 |
