siRNA Details
Gene Symbol: PRAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032391.2 | NP_115767.1 | 2 | 293 | ||
GenBank mRNA | AF331165.1 | - | 2 | 293 | ||
BC030950.1 | - | 2 | 291 | 4739311 | ||
BU535394.1 | - | 2 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
