siRNA Details
Gene Symbol: PRDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135610.1 | NP_001129082.1 | 3 | 218 | ||
NM_012231.4 | NP_036363.2 | 3 | 948 | |||
NM_015866.4 | NP_056950.2 | 3 | 948 | |||
XM_017002253.1 | XP_016857742.1 | 2 | 170 | |||
XM_017002254.1 | XP_016857743.1 | 2 | 170 | |||
XM_017002255.1 | XP_016857744.1 | 2 | 180 | |||
XM_017002256.1 | XP_016857745.1 | 3 | 249 | |||
XM_017002257.1 | XP_016857746.1 | 2 | 178 | |||
XM_017002264.1 | XP_016857753.1 | 2 | 1509 | |||
GenBank mRNA | U17838.1 | - | 3 | 947 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
