siRNA Details
Gene Symbol: TFEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012252.3 | NP_036384.1 | 3 | 459 | ||
XM_011515963.1 | XP_011514265.1 | 5 | 646 | |||
XM_011515964.2 | XP_011514266.1 | 5 | 661 | |||
XM_011515965.2 | XP_011514267.1 | 5 | 601 | |||
XM_011515966.2 | XP_011514268.1 | 5 | 710 | |||
XM_011515969.1 | XP_011514271.1 | 5 | 646 | |||
XM_011515972.2 | XP_011514274.1 | 2 | 175 | |||
XM_017011875.1 | XP_016867364.1 | 4 | 597 | |||
XM_017011877.1 | XP_016867366.1 | 2 | 174 | |||
GenBank mRNA | AK301479.1 | - | 5 | 651 | ||
AK313546.1 | - | 3 | 411 | |||
D43945.1 | - | 3 | 388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
